siRNA Details
Gene Symbol: SPATA41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028139.1 | 2 | 2346 | |||
NR_028140.1 | 3 | 754 | ||||
GenBank mRNA | AK057536.1 | - | 2 | 1283 | ||
BC028144.1 | - | 3 | 750 | 5214282 | ||
BC132704.1 | - | 3 | 697 | |||
BC132706.1 | - | 3 | 697 |
