siRNA Details
Gene Symbol: CARD17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007232.1 | NP_001007233.1 | 4 | 431 | ||
GenBank mRNA | AY761064.1 | - | 4 | 431 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CARD17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007232.1 | NP_001007233.1 | 4 | 431 | ||
GenBank mRNA | AY761064.1 | - | 4 | 431 |