siRNA Details
Gene Symbol: ANKRD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019644.3 | NP_062618.2 | 3, 4 | 649 | ||
GenBank mRNA | AK302407.1 | - | 3, 4 | 603 | ||
BC007020.2 | - | 1, 2 | 183 | |||
BC032799.1 | - | 3, 4 | 410 | 5170634 | ||
D78334.1 | - | 3, 4 | 405 |
