siRNA Details
Gene Symbol: LOC149950 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034152.1 | 3 | 490 | |||
GenBank mRNA | BC111383.1 | - | 3 | 490 | 5169644 | |
BC130462.1 | - | 2 | 288 | 40146493 | ||
BC130464.1 | - | 2 | 288 | 40146495 |
