siRNA Details
Gene Symbol: BREA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015445.1 | 1 | 498 | |||
GenBank mRNA | AL833948.1 | - | 1 | 498 | ||
AY940073.1 | - | 1 | 1467 | |||
BC132838.1 | - | 1 | 324 | |||
BC132840.1 | - | 1 | 324 |
