siRNA Details
Gene Symbol: MTX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010891.4 | NP_001010891.4 | 8 | 2709 | ||
NM_001167741.1 | NP_001161213.1 | 8 | 2728 | |||
XM_005248495.3 | XP_005248552.1 | 9 | 2800 | |||
XM_017009440.1 | XP_016864929.1 | 7 | 2633 | |||
GenBank mRNA | BC035085.1 | - | 1 | 963 | ||
BC150295.1 | - | 8 | 2702 | |||
BX538064.1 | - | 8 | 2711 | |||
BX647596.1 | - | 6 | 1034 |
