siRNA Details
Gene Symbol: SLC2A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020041.2 | NP_064425.2 | 1 | 202 | ||
XM_006713968.3 | XP_006714031.1 | 1 | 314 | |||
XM_011513856.2 | XP_011512158.1 | 1 | 339 | |||
XM_011513859.2 | XP_011512161.1 | 1 | 309 | |||
XM_011513860.2 | XP_011512162.1 | 1 | 309 | |||
XM_011513861.2 | XP_011512163.1 | 1 | 311 | |||
XM_011513862.2 | XP_011512164.1 | 1 | 347 | |||
XM_011513865.2 | XP_011512167.1 | 1 | 313 | |||
XM_011513866.2 | XP_011512168.1 | 1 | 311 | |||
XM_011513868.2 | XP_011512170.1 | 1 | 330 | |||
XM_017008457.1 | XP_016863946.1 | 1 | 327 | |||
XM_017008458.1 | XP_016863947.1 | 1 | 312 | |||
GenBank mRNA | AF210317.1 | - | 1 | 195 | ||
BC110414.1 | - | 1 | 232 | 5762360 | ||
BI821650.1 | - | 1 | 206 | |||
CD629819.1 | - | 1 | 207 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
