siRNA Details
Gene Symbol: SFMBT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018039.1 | NP_001018049.1 | 15, 16 | 1797 | ||
NM_001029880.2 | NP_001025051.1 | 15, 16 | 1899 | |||
XM_005252551.2 | XP_005252608.1 | 15, 16 | 1680 | |||
XM_006717490.2 | XP_006717553.1 | 13, 14 | 1672 | |||
XM_011519611.1 | XP_011517913.1 | 15, 16 | 1898 | |||
XM_017016466.1 | XP_016871955.1 | 15, 16 | 1800 | |||
GenBank mRNA | AB046837.1 | - | 15, 16 | 1797 | ||
BC152430.1 | - | 15, 16 | 1797 | 8862546 |
