siRNA Details
Gene Symbol: ZNF609 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015042.1 | NP_055857.1 | 9 | 8435 | ||
XM_011521386.2 | XP_011519688.1 | 10 | 8661 | |||
GenBank mRNA | AB002293.1 | - | 6 | 7146 | ||
BC050315.1 | - | 1 | 4052 |
