siRNA Details
Gene Symbol: C1GALT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020156.4 | NP_064541.1 | 2 | 333 | ||
XM_005249812.1 | XP_005249869.1 | 2 | 401 | |||
XM_011515453.2 | XP_011513755.1 | 3 | 530 | |||
XM_011515455.2 | XP_011513757.1 | 3 | 452 | |||
XM_011515456.2 | XP_011513758.1 | 2 | 264 | |||
XM_017012442.1 | XP_016867931.1 | 3 | 452 | |||
XM_017012443.1 | XP_016867932.1 | 3 | 447 | |||
XM_017012444.1 | XP_016867933.1 | 4 | 438 | |||
XM_017012445.1 | XP_016867934.1 | 5 | 653 | |||
XM_017012446.1 | XP_016867935.1 | 2 | 282 | |||
XM_017012447.1 | XP_016867936.1 | 3 | 376 | |||
XM_017012448.1 | XP_016867937.1 | 2 | 287 | |||
GenBank mRNA | AF155582.1 | - | 1 | 172 | ||
AJ132443.1 | - | 1 | 110 | |||
AJ243256.1 | - | 1 | 110 | |||
AJ278960.1 | - | 1 | 110 | |||
AK309328.1 | - | 2 | 318 | |||
DA977378.1 | - | 2 | 333 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
