siRNA Details
Gene Symbol: Rere | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053885.2 | NP_446337.2 | 23 | 5322 | ||
XM_006239438.2 | XP_006239500.1 | 23 | 5416 | |||
XM_006239441.2 | XP_006239503.1 | 17 | 7326 | |||
XM_008764198.1 | XP_008762420.1 | 23 | 5419 | |||
XM_008764199.1 | XP_008762421.1 | 23 | 5836 | |||
XM_008764200.1 | XP_008762422.1 | 23 | 5392 | |||
XM_008764201.1 | XP_008762423.1 | 22 | 5294 | |||
XM_008764202.1 | XP_008762424.1 | 22 | 4860 | |||
XM_008764203.1 | XP_008762425.1 | 21 | 6987 | |||
GenBank mRNA | CA511326.1 | - | 4 | 476 | ||
U44091.1 | - | 22 | 6131 |
