siRNA Details
Gene Symbol: Olr954 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000703.1 | NP_001000703.1 | 295 |
Gene Symbol: LOC100911571 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765074.1 | XP_008763296.1 | 1 | 295 |
Gene Symbol: LOC103692768 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765076.1 | XP_008763298.1 | 1 | 295 |
