siRNA Details
Gene Symbol: SERTAD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019605.3 | NP_062551.1 | 3 | 442 | ||
XM_005273193.4 | XP_005273250.1 | 3 | 642 | |||
GenBank mRNA | AK096381.1 | - | 3 | 408 | ||
AK315386.1 | - | 2 | 441 | |||
BC012083.1 | - | 3 | 442 | 4564813 |
