siRNA Details
Gene Symbol: SLCO1B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019844.3 | NP_062818.1 | 9 | 999 | ||
GenBank mRNA | AF187815.1 | - | 7 | 758 | ||
AJ251506.1 | - | 7 | 802 | |||
AK055874.1 | - | 9 | 977 | |||
AY342017.1 | - | 7 | 772 | |||
BC022843.1 | - | 8 | 883 | |||
BC058006.1 | - | 8 | 888 | |||
BC070264.1 | - | 8 | 862 | |||
BC105597.1 | - | 8 | 888 |
Gene Symbol: SLCO1B7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009562.4 | NP_001009562.3 | 6 | 617 | ||
GenBank mRNA | AF401642.1 | - | 6 | 617 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
