siRNA Details
Gene Symbol: FHL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039492.2 | NP_001034581.1 | 3, 4 | 388 | ||
NM_001318894.1 | NP_001305823.1 | 2, 3 | 634 | |||
NM_001318895.1 | NP_001305824.1 | 3, 4 | 622 | |||
NM_001318896.1 | NP_001305825.1 | 2, 3 | 337 | |||
NM_001318899.1 | NP_001305828.1 | 4, 5 | 510 | |||
NM_001450.3 | NP_001441.4 | 2, 3 | 571 | |||
NM_201555.1 | NP_963849.1 | 4, 5 | 743 | |||
NM_201557.3 | NP_963851.2 | 2, 3 | 563 | |||
XM_011510798.2 | XP_011509100.1 | 3, 4 | 370 | |||
GenBank mRNA | AK313841.1 | - | 3, 4 | 420 | ||
AL521653.3 | - | 2, 3 | 264 | |||
AY634685.1 | - | 1, 2 | 164 | |||
BC012742.2 | - | 4, 5 | 743 | 3840517 | ||
BC014397.1 | - | 4, 5 | 743 | 3546686 | ||
BC093049.1 | - | 2, 3 | 329 | 30374919 | ||
BM559077.1 | - | 4, 5 | 392 | |||
BM836767.1 | - | 2, 3 | 337 | |||
BP257466.1 | - | 2, 3 | 564 | |||
BT006960.1 | - | 1, 2 | 164 | |||
CF552311.1 | - | 2, 3 | 294 | |||
CR457437.1 | - | 1, 2 | 164 | |||
CR936799.1 | - | 2, 3 | 339 | |||
DA682076.1 | - | 3, 4 | 499 | |||
DQ266432.1 | - | 3, 4 | 622 | |||
DQ307067.1 | - | 2, 3 | 501 | |||
JQ313902.1 | - | 1, 2 | 134 | |||
L42176.1 | - | 3, 4 | 302 | |||
U29332.1 | - | 2, 3 | 285 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
