siRNA Details
Gene Symbol: Rbms2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025403.1 | NP_001020574.1 | 12 | 1180 | ||
XM_006240742.2 | XP_006240804.1 | 16 | 1413 | |||
XM_006240744.2 | XP_006240806.1 | 14 | 1582 | |||
XM_008765021.1 | XP_008763243.1 | 15 | 1669 | |||
XM_008765022.1 | XP_008763244.1 | 14 | 1589 | |||
XM_008765023.1 | XP_008763245.1 | 13 | 1406 | |||
GenBank mRNA | BC097377.1 | - | 12 | 1180 | 7445017 |
