siRNA Details
Gene Symbol: Nars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025635.2 | NP_001020806.2 | 9 | 795 | ||
NM_001142949.1 | NP_001136421.1 | 7 | 642 | |||
XM_006254844.2 | XP_006254906.1 | 9 | 853 | |||
GenBank mRNA | AY387099.1 | - | 6 | 548 | ||
BC099099.1 | - | 8 | 676 | 7459693 |
