siRNA Details
Gene Symbol: Dusp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025650.1 | NP_001020821.1 | 9 | 1111 | ||
GenBank mRNA | BC098712.1 | - | 9 | 1111 | 7321787 |
Gene Symbol: LOC103690006 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763101.1 | XP_008761323.1 | 9 | 1190 |
