siRNA Details
Gene Symbol: Rad23b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025275.1 | NP_001020446.1 | 10 | 2730 | ||
GenBank mRNA | BC090351.1 | - | 7 | 2193 | ||
BC098674.1 | - | 10 | 2730 | 7190031 | ||
BC111406.1 | - | 5 | 1869 |
