siRNA Details
Gene Symbol: Lrrc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025659.2 | NP_001020830.2 | 4 | 568 | ||
GenBank mRNA | BC097394.1 | - | 4 | 568 | 7453303 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lrrc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025659.2 | NP_001020830.2 | 4 | 568 | ||
GenBank mRNA | BC097394.1 | - | 4 | 568 | 7453303 |