siRNA Details
Gene Symbol: Ei24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025660.1 | NP_001020831.1 | 12 | 1650 | ||
XM_006242800.2 | XP_006242862.1 | 11 | 1741 | |||
XM_006242801.2 | XP_006242863.1 | 11 | 1698 | |||
GenBank mRNA | BC098721.1 | - | 12 | 1650 | 7370909 |
