siRNA Details
Gene Symbol: Pus1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025563.1 | NP_001020734.1 | 5 | 914 | ||
XM_006249547.2 | XP_006249609.1 | 5 | 1051 | |||
XM_008769330.1 | XP_008767552.1 | 5 | 1059 | |||
XM_008769331.1 | XP_008767553.1 | 5 | 1055 | |||
GenBank mRNA | BC161821.1 | - | 5 | 874 | 9027947 |
