siRNA Details
Gene Symbol: Mettl18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025668.1 | NP_001020839.1 | 2 | 641 | ||
XM_006250155.2 | XP_006250217.1 | 2 | 568 | |||
GenBank mRNA | BC098702.1 | - | 2 | 641 | 7388665 |
