siRNA Details
Gene Symbol: Rassf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025671.1 | NP_001020842.1 | 10 | 1829 | ||
XM_006250745.2 | XP_006250807.1 | 11 | 1941 | |||
XM_006250747.2 | XP_006250809.1 | 11 | 1929 | |||
XM_006250748.1 | XP_006250810.1 | 11 | 1964 | |||
XM_008770036.1 | XP_008768258.1 | 11 | 2024 | |||
XM_008770037.1 | XP_008768259.1 | 11 | 1974 | |||
XM_008770038.1 | XP_008768260.1 | 11 | 5216 | |||
GenBank mRNA | BC097375.1 | - | 10 | 1829 | 7444629 |
