siRNA Details
Gene Symbol: Ttc23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025681.1 | NP_001020852.2 | 10 | 1313 | ||
XM_008759515.1 | XP_008757737.1 | 11 | 2247 | |||
XM_008759516.1 | XP_008757738.1 | 11 | 1774 | |||
GenBank mRNA | BC097953.1 | - | 10 | 1313 | 7455883 |

More Information
Other Species Targeted

