siRNA Details
Gene Symbol: RGD1311739 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025691.1 | NP_001020862.1 | 6 | 1184 | ||
GenBank mRNA | BC098814.1 | - | 6 | 1184 | 7377135 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RGD1311739 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025691.1 | NP_001020862.1 | 6 | 1184 | ||
GenBank mRNA | BC098814.1 | - | 6 | 1184 | 7377135 |