siRNA Details
Gene Symbol: Cops5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025695.1 | NP_001020866.1 | 7 | 1005 | ||
XM_006237758.2 | XP_006237820.1 | 7 | 945 | |||
GenBank mRNA | BC098736.1 | - | 7 | 1005 | 7385252 | |
FQ214411.1 | - | 7 | 928 | |||
FQ215612.1 | - | 7 | 928 | |||
FQ227775.1 | - | 7 | 929 | |||
FQ232542.1 | - | 7 | 928 |
