siRNA Details
Gene Symbol: Hsdl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025697.1 | NP_001020868.1 | 11 | 1668 | ||
XM_008763735.1 | XP_008761957.1 | 12 | 1603 | |||
XM_008763736.1 | XP_008761958.1 | 12 | 1554 | |||
GenBank mRNA | BC097407.1 | - | 11 | 1668 | 7454038 |
