siRNA Details
Gene Symbol: Tmem57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025699.2 | NP_001020870.2 | 4 | 595 | ||
XM_008764240.1 | XP_008762462.1 | 3 | 791 | |||
XM_008764241.1 | XP_008762463.1 | 4 | 547 | |||
GenBank mRNA | AY845021.1 | - | 4 | 595 | ||
BC097999.1 | - | 4 | 521 | 7453136 |
