siRNA Details
Gene Symbol: Celf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025421.1 | NP_001020592.1 | 13 | 1837 | ||
XM_006234513.2 | XP_006234575.1 | 14 | 5888 | |||
XM_006234514.1 | XP_006234576.1 | 14 | 5010 | |||
XM_006234516.1 | XP_006234578.1 | 14 | 1978 | |||
XM_006234518.1 | XP_006234580.1 | 14 | 1975 | |||
XM_006234519.1 | XP_006234581.1 | 13 | 5086 | |||
XM_006234520.1 | XP_006234582.1 | 13 | 5083 | |||
XM_006234521.1 | XP_006234583.1 | 14 | 4995 | |||
XM_006234522.1 | XP_006234584.1 | 13 | 5020 | |||
XM_006234523.1 | XP_006234585.1 | 14 | 5156 | |||
XM_006234524.2 | XP_006234586.1 | 14 | 5047 | |||
XM_006234526.2 | XP_006234588.1 | 14 | 5271 | |||
XM_006234527.2 | XP_006234589.1 | 14 | 5268 | |||
XM_008762008.1 | XP_008760230.1 | 15 | 6090 | |||
XM_008762009.1 | XP_008760231.1 | 15 | 5173 | |||
XM_008762010.1 | XP_008760232.1 | 14 | 5937 | |||
XM_008762011.1 | XP_008760233.1 | 13 | 4967 | |||
GenBank mRNA | BC098012.1 | - | 13 | 1837 | 7454358 | |
FQ222063.1 | - | 14 | 1976 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001244903, NM_017368, NM_198683, NM_001244891.
Gene Ontology Categories:
