siRNA Details
Gene Symbol: Procr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025733.2 | NP_001020904.1 | 3 | 681 | ||
GenBank mRNA | BC097380.1 | - | 3 | 652 | 7445753 | |
BC099115.1 | - | 3 | 681 | 7462042 | ||
FQ220150.1 | - | 3 | 720 | |||
FQ221148.1 | - | 3 | 688 | |||
FQ228484.1 | - | 3 | 686 | |||
FQ228710.1 | - | 3 | 688 | |||
FQ229445.1 | - | 3 | 718 | |||
FQ229483.1 | - | 3 | 688 |
