siRNA Details
Gene Symbol: Adhfe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025423.1 | NP_001020594.1 | 6, 7 | 572 | ||
XM_008763507.1 | XP_008761729.1 | 6, 7 | 953 | |||
XM_008763508.1 | XP_008761730.1 | 7, 8 | 620 | |||
XM_008763509.1 | XP_008761731.1 | 4, 5 | 374 | |||
XM_008763510.1 | XP_008761732.1 | 4, 5 | 751 | |||
GenBank mRNA | BC085821.1 | - | 6, 7 | 575 | ||
BC097945.1 | - | 6, 7 | 572 | 7323001 |
