siRNA Details
Gene Symbol: Adhfe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025423.1 | NP_001020594.1 | 9 | 862 | ||
XM_008763507.1 | XP_008761729.1 | 9 | 1243 | |||
XM_008763508.1 | XP_008761730.1 | 10 | 910 | |||
XM_008763509.1 | XP_008761731.1 | 7 | 664 | |||
XM_008763510.1 | XP_008761732.1 | 7 | 1041 | |||
GenBank mRNA | BC085821.1 | - | 9 | 865 | ||
BC097945.1 | - | 9 | 862 | 7323001 |
