siRNA Details
Gene Symbol: Adhfe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025423.1 | NP_001020594.1 | 3 | 146 | ||
XM_008763507.1 | XP_008761729.1 | 3 | 527 | |||
XM_008763508.1 | XP_008761730.1 | 4 | 194 | |||
GenBank mRNA | BC085821.1 | - | 3 | 149 | ||
BC097945.1 | - | 3 | 146 | 7323001 |
