siRNA Details
Gene Symbol: Wdr61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025743.1 | NP_001020914.1 | 11 | 1070 | ||
XM_006243085.2 | XP_006243147.1 | 11 | 1071 | |||
XM_006243086.2 | XP_006243148.1 | 11 | 1124 | |||
XM_006243087.2 | XP_006243149.1 | 11 | 1097 | |||
GenBank mRNA | BC098059.1 | - | 11 | 1070 | 7458076 |
