siRNA Details
Gene Symbol: Tmem186 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025756.1 | NP_001020927.1 | 2 | 1062 | ||
GenBank mRNA | BC098933.1 | - | 2 | 1062 | 7386881 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmem186 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025756.1 | NP_001020927.1 | 2 | 1062 | ||
GenBank mRNA | BC098933.1 | - | 2 | 1062 | 7386881 |