siRNA Details
Gene Symbol: MGC112692 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025759.1 | NP_001020930.1 | 729 | |||
XM_008768997.1 | XP_008767219.1 | 2 | 765 | |||
GenBank mRNA | BC081909.1 | - | 725 | |||
BC098706.1 | - | 729 | 7114401 |
Gene Symbol: LOC100910238 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008769043.1 | XP_008767265.1 | 350 |
Gene Symbol: LOC100910352 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008769042.1 | XP_008767264.1 | 350 |
