siRNA Details
Gene Symbol: Tjp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053773.1 | NP_446225.1 | 4 | 434 | ||
XM_006231175.2 | XP_006231237.1 | 4 | 1516 | |||
XM_006231176.2 | XP_006231238.1 | 4 | 1517 | |||
XM_006231177.2 | XP_006231239.1 | 3 | 747 | |||
XM_006231178.2 | XP_006231240.1 | 3 | 3676 | |||
XM_006231179.2 | XP_006231241.1 | 3 | 580 | |||
XM_006231181.2 | XP_006231243.1 | 4 | 535 | |||
XM_006231182.2 | XP_006231244.1 | 3 | 747 | |||
XM_008760302.1 | XP_008758524.1 | 4 | 276 | |||
GenBank mRNA | BC103479.1 | - | 4 | 434 | 7936785 |
