siRNA Details
Gene Symbol: Orc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033690.1 | NP_001028862.1 | 7 | 1121 | ||
XM_006255357.1 | XP_006255419.1 | 6 | 711 | |||
GenBank mRNA | BC101871.1 | - | 7 | 1121 | 5621693 |
