siRNA Details
Gene Symbol: Plcb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033350.1 | NP_203501.1 | 17 | 1949 | ||
XM_006230711.2 | XP_006230773.1 | 17 | 2081 | |||
GenBank mRNA | DQ120507.1 | - | 17 | 1949 | ||
DQ120508.1 | - | 17 | 1949 | |||
M99567.1 | - | 17 | 1883 |
