siRNA Details
Gene Symbol: Depdc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029916.1 | NP_001025087.1 | 7 | 1296 | ||
GenBank mRNA | BC097357.1 | - | 7 | 1296 | 7442188 |

More Information
Other Species Targeted

Gene Ontology Categories:
