siRNA Details
Gene Symbol: Ubox5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033997.1 | NP_001029169.1 | 6 | 2719 | ||
XM_008762160.1 | XP_008760382.1 | 5 | 2630 | |||
XM_008762161.1 | XP_008760383.1 | 4 | 2471 | |||
GenBank mRNA | BC101912.1 | - | 6 | 2719 | 7933511 |
