siRNA Details
Gene Symbol: Jph2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037974.1 | NP_001033063.1 | 4 | 1350 | ||
XM_006235526.2 | XP_006235588.1 | 4 | 2241 | |||
GenBank mRNA | DQ304564.1 | - | 4 | 1350 | ||
FQ223967.1 | - | 4 | 1777 |
