siRNA Details
Gene Symbol: Slc25a40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037186.1 | NP_001032263.1 | 1036 | |||
GenBank mRNA | BC100071.1 | - | 1036 | 7322585 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc25a40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037186.1 | NP_001032263.1 | 1036 | |||
GenBank mRNA | BC100071.1 | - | 1036 | 7322585 |