siRNA Details
Gene Symbol: L3mbtl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033695.1 | NP_001028867.1 | 17 | 2504 | ||
XM_008765711.1 | XP_008763933.1 | 17 | 2532 | |||
GenBank mRNA | BC088331.1 | - | 4 | 908 | ||
BC101865.1 | - | 17 | 2504 | 7938856 |
