siRNA Details
Gene Symbol: KCMF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020122.4 | NP_064507.3 | 7 | 1688 | ||
XM_006712052.3 | XP_006712115.1 | 7 | 1720 | |||
XM_011532990.1 | XP_011531292.1 | 7 | 1610 | |||
XM_017004511.1 | XP_016860000.1 | 6 | 1324 | |||
GenBank mRNA | AB083199.1 | - | 7 | 1397 | ||
AF155652.1 | - | 7 | 1397 | |||
AK023403.1 | - | 7 | 1367 | |||
AK311099.1 | - | 1 | 229 | |||
AL122115.1 | - | 5 | 1041 | |||
BC000178.2 | - | 7 | 1487 | 3048124 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_019715, AK215477, BC043330, BC029325, AK007070, and others.
Gene Ontology Categories:
