siRNA Details
Gene Symbol: Baiap2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034140.1 | NP_001029312.1 | 8 | 932 | ||
GenBank mRNA | BC089216.1 | - | 7 | 775 | ||
BC105815.1 | - | 8 | 932 | 7367101 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Baiap2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034140.1 | NP_001029312.1 | 8 | 932 | ||
GenBank mRNA | BC089216.1 | - | 7 | 775 | ||
BC105815.1 | - | 8 | 932 | 7367101 |