siRNA Details
Gene Symbol: Tmem177 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037776.1 | NP_001032865.1 | 2 | 263 | ||
XM_006249664.2 | XP_006249726.1 | 2 | 510 | |||
XM_006249665.2 | XP_006249727.1 | 3 | 402 | |||
GenBank mRNA | BC098688.1 | - | 2 | 263 | 7386020 |
