siRNA Details
Gene Symbol: Rpgrip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006251921.2 | XP_006251983.1 | 12 | 1382 | ||
XM_008770631.1 | XP_008768853.1 | 12 | 1350 | |||
XM_008770632.1 | XP_008768854.1 | 12 | 1380 | |||
XM_008770633.1 | XP_008768855.1 | 11 | 1252 | |||
XM_008770634.1 | XP_008768856.1 | 12 | 1379 | |||
XM_008770635.1 | XP_008768857.1 | 12 | 1375 | |||
XM_008770636.1 | XP_008768858.1 | 12 | 1375 | |||
GenBank mRNA | BC097397.1 | - | 2 | 292 |
