siRNA Details
Gene Symbol: GRIPAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020137.4 | NP_064522.3 | 26 | 2777 | ||
XM_011543935.1 | XP_011542237.1 | 26 | 2807 | |||
XM_011543936.1 | XP_011542238.1 | 25 | 2729 | |||
XM_011543937.1 | XP_011542239.1 | 26 | 2854 | |||
XM_017029656.1 | XP_016885145.1 | 25 | 2699 | |||
XM_017029657.1 | XP_016885146.1 | 26 | 3178 | |||
XM_017029658.1 | XP_016885147.1 | 26 | 4087 | |||
XM_017029659.1 | XP_016885148.1 | 26 | 2824 | |||
XM_017029660.1 | XP_016885149.1 | 25 | 3100 | |||
XM_017029661.1 | XP_016885150.1 | 25 | 2746 | |||
GenBank mRNA | AB032993.1 | - | 26 | 2722 | ||
AK292158.1 | - | 26 | 2748 | |||
AL136847.1 | - | 13 | 1663 | |||
BC001522.2 | - | 16 | 1890 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
